missing translation for 'onlineSavingsMsg'
Learn More

GFAP Antibody, Novus Biologicals™

Product Code. 18243761 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
Unit Size:
0.25mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18243761 0.25 mL 0.25mL
18229130 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18243761 Supplier Novus Biologicals Supplier No. NB120169970.25ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GFAP Polyclonal specifically detects GFAP in Human, Mouse, Rat, Bovine, Chicken, Feline, Guinea Pig, Primate, Sheep samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:250
Formulation Citrate, pH 6.0, containing less than 0.1% sodium azide with Sodium Azide
Gene Alias FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Gene Symbols GFAP
Host Species Rabbit
Immunogen Full length native protein (purified) (Cow). Glial fibrillary protein isolated from cow spinal cord
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cytoskeleton Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Test Specificity GFAP
Target Species Feline, Human, Guinea Pig, Bovine, Primate, Mouse, Sheep, Chicken, Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.