missing translation for 'onlineSavingsMsg'
Learn More

GFAP Antibody, Novus Biologicals™

Product Code. 18229130 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
Unit Size:
0.25mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18229130 1 mL 1mL
18243761 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18229130 Supplier Novus Biologicals Supplier No. NB12016997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GFAP Polyclonal antibody specifically detects GFAP in Human, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:250, Immunohistochemistry-Paraffin 1:50-1:250
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Alias FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Gene Symbols GFAP
Host Species Rabbit
Immunogen This GFAP Antibody was developed against glial fibrillary protein isolated from cow spinal cord.
Molecular Weight of Antigen 50 kDa
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Test Specificity GFAP
Target Species Human, Bovine, Rat, Chicken, Feline, Guinea Pig, Primate, Sheep
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.