missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephacryl™ High Resolution Media
Purify proteins, polysaccharides, and other macromolecules by gel filtration at laboratory and industrial scale. Cytiva Life Sciences™Sephacryl™ High Resolution Media enables fast and reproducible purifications.
£166.00 - £676.00
Specifications
Buffer | 0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place) |
---|---|
Height (English) | 5.9 in. |
Height (Metric) | 15cm |
Max. Pressure | 1 bar |
pH Range | 3 to 11 |
Product Code | Brand | Product Type | Quantity | Flow Rate | Particle Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Flow Rate | Particle Size | Price | Quantity & Availability | ||
11530794
|
Cytiva
17-0612-01 |
Sephacryl S-100 HR media | 750mL | >125cm/hr. | 25 to 75μm |
£676.00
750mL |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
11510794
|
Cytiva
17-0609-01 |
Sephacryl S-400 HR media | 750mL | >150cm/hr. | 50μm |
£676.00
750mL |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
11500794
|
Cytiva
17-0599-10 |
Sephacryl S-300 HR media | 150mL | >150cm/hr. | 50μm |
£166.00
150mL |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
11580784
|
Cytiva
17-0584-10 |
Sephacryl S-200 HR media | 150mL | >150cm/hr. | 50μm |
£166.00
150mL |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
Description
- Fast and reproducible purification of proteins, polysaccharides, and other macromolecules
- Gel filtration media providing high resolution with fast and easy operation
- Available in five different fractionation ranges, allows for reproducible separations over a wide molecular weight range
- BioProcess medium supported for industrial applications and well-established in approved processes
- Available in different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
- Long-term physical, chemical stability and easy handling
- Hydrophilic nature enables maximum recovery and minimum nonspecific adsorption
- Average particle Size: 50μm
- Matrix: Allyl dextran and N, N'-methylenebisacrylamide
- pH stability working range: 3-11
- Shelf Life: 5years
Specifications
0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place) | |
15cm | |
3 to 11 | |
4° to 30°C | |
Gel Filtration Media |
5.9 in. | |
1 bar | |
Allyl Dextran and N, N'-methylenebisacrylamide | |
Purification of Proteins, Polysaccharides and other macromolecules |