missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephacryl™ High Resolution Media

Artikelnummer. 11580784 Alle ansehen Cytiva Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
150 mL
750 mL
Product Type:
Sephacryl S-100 HR media
Sephacryl S-200 HR media
Sephacryl S-300 HR media
Sephacryl S-400 HR media
Flow Rate:
>125 cm/hr.
>150 cm/hr.
Particle Size:
25 to 75μm
50μm
missing translation for 'unitSize'
150mL
750mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11580784

missing translation for 'mfr': Cytiva 17058410

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Purify proteins, polysaccharides, and other macromolecules by gel filtration at laboratory and industrial scale. Cytiva Life Sciences™Sephacryl™ High Resolution Media enables fast and reproducible purifications.

  • Fast and reproducible purification of proteins, polysaccharides, and other macromolecules
  • Gel filtration media providing high resolution with fast and easy operation
  • Available in five different fractionation ranges, allows for reproducible separations over a wide molecular weight range
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Available in different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Long-term physical, chemical stability and easy handling
  • Hydrophilic nature enables maximum recovery and minimum     nonspecific adsorption

  • Average particle Size: 50μm
  • Matrix: Allyl dextran and N, N'-methylenebisacrylamide
  • pH stability working range: 3-11
  • Shelf Life: 5years

TRUSTED_SUSTAINABILITY

Spezifikation

Buffer 0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place)
Product Type Sephacryl S-200 HR media
Quantity 150 mL
Height (English) 5.9 in.
Height (Metric) 15cm
Flow Rate >150 cm/hr.
Max. Pressure 1 bar
pH Range 3 to 11
Particle Size 50μm
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Content And Storage 4° to 30°C
For Use With (Application) Purification of Proteins, Polysaccharides and other macromolecules
Matrix Gel Filtration Media
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Sephacryl S-200 High Resolution, 150 ml
Signalwort
  • Warning
Gefahrenkategorie
  • Flammable liquid Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contains : ethanol
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt