missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCDFP-15 Mouse, Clone: PIP/1571, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | GCDFP-15 |
---|---|
Clone | PIP/1571 |
Concentration | 200μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940648
|
EnquireBio
5304-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930648
|
EnquireBio
5304-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
GCDFP-15 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
5304 | |
Recombinant human GCDFP-15 protein fragment (aa 41-146) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
15kDa |
PIP/1571 | |
Western Blot | |
Unconjugated | |
RUO | |
NP_002643 | |
PIP | |
IgG2a κ | |
Protein A/G Purified | |
Cytoplasmic (Golgi bodies) and Secreted | |
HepG2 Cells. Breast or Pancreas. |