missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCDFP-15 Mouse, Clone: PIP/1571, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 5304-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
GCDFP-15 | |
Cytoplasmic (Golgi bodies) and Secreted | |
PIP/1571 | |
Unconjugated | |
NP_002643 | |
Mouse | |
15kDa | |
Protein A/G Purified | |
RUO | |
5304 | |
IgG2a κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
PIP | |
Recombinant human GCDFP-15 protein fragment (aa 41-146) | |
HepG2 Cells. Breast or Pancreas. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |