missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 2 Rabbit anti-Human, Mouse, Rat, Clone: 6G9O8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | GABA-A R beta 2 |
|---|---|
| Clone | 6G9O8 |
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18353445
|
Novus Biologicals
NBP3-15403-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375855
|
Novus Biologicals
NBP3-15403-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA-A R beta 2 Monoclonal antibody specifically detects GABA-A R beta 2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GABA-A R beta 2 | |
| Western Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GABA(A) receptor subunit beta-2, gamma-aminobutyric acid (GABA) A receptor, beta 2, gamma-aminobutyric acid A receptor beta 2, gamma-aminobutyric acid receptor subunit beta-2, MGC119386, MGC119388, MGC119389 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human GABA-A R beta 2 (GABRB2) (P47870).PYVKAIDMYLMGCFVFVFMALLEYALVNYIFFGRGPQRQKKAAEKAASANNEKMRLDVNKIFYKDIKQNGTQYRSLWDPTGNLSPTRRTTNYDFSLYTMDP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 6G9O8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2561 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title