missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, 97+%, Thermo Scientific™

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

£59.70 - £203.00

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.739
MDL Number MFCD00010549
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
Synonym furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
PubChem CID 3440
ChEBI CHEBI:47426
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
SMILES C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404059
SDS View Documents
Thermo Scientific Alfa Aesar
J61457.06
5g
£59.70
5g
View alternative products
Save Up to 
»
Estimated Shipment: 07-10-2026
Add to basket
 
 Restricted Item
15414059
SDS View Documents
Thermo Scientific Alfa Aesar
J61457.09
10g
£106.00
10g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15424059
SDS View Documents
Thermo Scientific Alfa Aesar
J61457.14
25g
£203.00
25g
View alternative products
Save Up to 
»
Estimated Shipment: 11-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

54-31-9
330.739
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
3440
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
C12H11ClN2O5S
MFCD00010549
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
CHEBI:47426
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl

Specifications

54-31-9
MFCD00010549
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
3440
330.74
Furosemide
C12H11ClN2O5S
14,4309
Soluble in acetone, DMF or methanol; Slightly soluble in water
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
330.739
CHEBI:47426
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360

missing translation for 'einecsNumber' : 200-203-6

missing translation for 'rtecsNumber' : CB2625000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.