missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, 97+%, Thermo Scientific™

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

Brand:  Thermo Scientific Alfa Aesar J61457.09

Quantity 10g

 View more versions of this product

Product Code. 15414059

  • £106.00 / 10g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

54-31-9
330.739
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
3440
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
C12H11ClN2O5S
MFCD00010549
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
CHEBI:47426
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl

Specifications

54-31-9
MFCD00010549
14,4309
Soluble in acetone, DMF or methanol; Slightly soluble in water
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
330.739
CHEBI:47426
≥97%
C12H11ClN2O5S
10g
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
3440
330.74
Furosemide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.