missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, 97+%

Product Code. 15424059
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424059 25 g 25g
15404059 5 g 5g
15414059 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15424059 Supplier Thermo Scientific Alfa Aesar Supplier No. J61457.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter | CAS: 54-31-9 | C12H11ClN2O5S | 330.739 g/mol

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.739
MDL Number MFCD00010549
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
Synonym furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
PubChem CID 3440
ChEBI CHEBI:47426
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
SMILES C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl

Specifications

Melting Point 206°C to 209°C
Color White
Quantity 25 g
Merck Index 14,4309
Solubility Information Soluble in acetone, DMF or methanol; Slightly soluble in water
Formula Weight 330.74
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material Furosemide
compliance-icons
Product Identifier
  • Furosemide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Furosemide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.