missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB2 Mouse anti-Human, Clone: PCRP-FOXB2-2B2, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | FOXB2 |
|---|---|
| Clone | PCRP-FOXB2-2B2 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18317355
|
Novus Biologicals
NBP3-13972-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18310745
|
Novus Biologicals
NBP3-13972-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein ArraySpecifications
| FOXB2 | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q5VYV0 | |
| 442425 | |
| IgG2a | |
| Protein A or G purified |
| PCRP-FOXB2-2B2 | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| bA159H20.4, forkhead box B2, forkhead box protein B2 | |
| Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title