missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB2 Mouse anti-Human, Clone: PCRP-FOXB2-2B2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-13972-20UG
This item is not returnable.
View return policy
Description
FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein Array
Specifications
| FOXB2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Q5VYV0 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 442425 | |
| Store at 4°C. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array | |
| PCRP-FOXB2-2B2 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| bA159H20.4, forkhead box B2, forkhead box protein B2 | |
| Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction