missing translation for 'onlineSavingsMsg'
Learn More

FOXB2 Mouse anti-Human, Clone: PCRP-FOXB2-2B2, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-13972-100UG

 View more versions of this product

Product Code. 18310745

  • £396.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXB2
Monoclonal
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Q5VYV0
Mouse
Protein A or G purified
RUO
442425
Store at 4°C.
IgG2a
Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array
PCRP-FOXB2-2B2
Unconjugated
10 mM PBS with 0.05% BSA
bA159H20.4, forkhead box B2, forkhead box protein B2
Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0)
100 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.