missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FLT3, His Tag Recombinant Protein
FLT3 Recombinant Human Protein
£434.00 - £6900.00
Specifications
Accession Number | NP_004110 |
---|---|
Gene Alias | FLT3, FLK2, STK1, CD135 |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11539336
|
Invitrogen
PV3182 |
10μg |
£434.00
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468413
|
Invitrogen
PR4666C |
100μg |
£1600.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348052
|
Invitrogen
PV4108 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.Specifications
NP_004110 | |
Human | |
Baculovirus |
FLT3, FLK2, STK1, CD135 | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.