missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FLT3, His Tag Recombinant Protein
FLT3 Recombinant Human Protein
Brand: Invitrogen PR4666C
Code : NEW
Additional Details : Weight : 0.01000kg
Description
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.Specifications
NP_004110 | |
48.6 kDa | |
FLT3, FLK2, STK1, CD135 | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
FLT-3 Ligand |
Kinase Assay | |
100μg | |
PDGFR Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.