missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FLT3, His Tag Recombinant Protein
FLT3 Recombinant Human Protein
Brand: Invitrogen PV4108
Code : 50
Additional Details : Weight : 0.05250kg
Description
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.Specifications
NP_004110 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
48.6kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
FLT3, FLK2, STK1, CD135 | |
FLT3 was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% NP-40, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 564-958 | |
2430nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Frozen |
0.35mg/mL | |
Frozen | |
FLT3 Recombinant Human Protein | |
–80°C | |
1 Each | |
FLT3 Recombinant Human Protein | |
PDGFR | |
FLT3 Recombinant Human Protein | |
Human | |
Baculovirus | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.