Learn More
Fisherbrand™ Fisherbrand Stainless Steel Forceps
Product Code. 15576579 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
160 mm
Product Code. Length (Metric)
15576579 160 mm
1 options
Product Code. 15576579
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Has straight, sharp ends, and smooth jaws
  • 160 mm Stainless Steel Forceps
  • Smooth jaws, particularly suitable for tissue handling samples or other non-marking applications
  • Supplied in standard pack of 1
  • Stainless steel is quick to clean and has an attractive shiny appearance making it the material of choice for applications requiring sterile surfaces
  • It is immune to sterilising solutions, most of the organic chemicals and dyestuffs and a wide variety of inorganic chemicals
  • Extremely durable
Specifications
Length (Metric) 160 mm
Material Stainless Steel
Quantity 1 Ea.
Style Straight, blunt ends, smooth jaws