missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Scoopula™ Spatula

Product Code. 16448115 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
15 cm
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
16448115 15 cm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16448115

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Dig out materials, core, and stir easily

  • Minimizes spilling in handling small amounts of material
  • More versatile
  • One pointed end, one blunt end
  • Stainless-steel
  • Does not rust

Specifications

Shape Pointed End, Blunt End
Product Type Scoopula™
For Use With (Application) Minimizes spilling in handling small amounts of material, dig out materials, core and stir easily
Length (English) 6 in.
Includes One pointed end and one blunt end
Material Stainless Steel
Length (Metric) 15 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.