missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Disposable Beakers

Product Code. p-6480429 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
15 mL
30 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
Unit Size:
Pack of 100
Pack of 25
Pack of 50

Product Code. 17445133

Only null left
Add to basket
Add to basket

Highly resistant to strong acids, organic solvents and other common laboratory reagents

Rib construction permits stacking without binding

  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.

Specifications

Capacity (Metric) 15 mL
Material Polypropylene
For Use With (Application) Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
Includes Spout
Increments 5 mL
Type Beaker
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.