missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Disposable Beakers

Product Code. 17485123 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
Unit Size:
Pack of 100
Pack of 25
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17485123 50 mL Pack of 100
17445133 15 mL Pack of 100
17455133 30 mL Pack of 100
17435133 100 mL Pack of 100
17495123 150 mL Pack of 100
17415133 250 mL Pack of 50
17405133 400 mL Pack of 50
17425133 600 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17485123
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket

Highly resistant to strong acids, organic solvents and other common laboratory reagents

Rib construction permits stacking without binding

  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.

Specifications

Capacity (Metric) 50 mL
Material Polypropylene
For Use With (Application) Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
Includes Spout
Increments 5 mL
Type Beaker
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.