Description
- Clear polycarbonate lens
- Soft, flexible PVC body molds to facial contours for a comfortable fit
- Adjustable headstrap ensures custom fit
- Resist a broad range of chemicals
- Circular vents allow air to flow freely through goggle body
Compliance
Meets ANSI/ISEA Z87.1-2015 American National Standard for Occupational and Educational Personal Eye and Face Protection Devices, Section 8.1 Droplet and Splash Hazard (D3)
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA Z87.1-2015 |
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Chemical Splash Protection Goggles |
| Material | Polycarbonate |
| Frame Material | PVC |