missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Traceable™ Bench Conductivity Meter measures conductivity, resistivity, dissolved solids, salinity, concentration, and temperature; includes certificate of traceability to standards provided by NIST™.
- Features six modes — conductivity, resistivity, total dissolved solids, salinity, concentration and temperature readings
- Ideal for ISO 9000 and government certification requirements
- Autorange automatically selects most appropriate range for current reading unless feature is disabled
- Controls allow operator to calibrate to solution standards; four calibration points ensure accuracy over entire range
- Automatic/manual temperature compensation with readings referenced to international standard of 25°C
- Temperature compensation disable function fulfills USP-NF (United States Pharmacopeia, National Formulary, 645 Conductivity Measurement) requirements
- Measures conductivity in chromatography, biology, food, electronics and water analysis
- Meets CAP, ASTM™, NCCLS, and ACS specifications in measuring reagent-grade water
- Includes certificate to indicate instrument traceability to standards provided by the National Institute of Standards and Technology; calibrated by an ISO 17025 calibration laboratory accredited by A2LA
- RS-232-C output allows readings to be downloaded to accessory datalogger or computer
- Conductivity Autoranging: 0.01 μS to 200 mS (in 5 ranges)
- Resistance Autoranging: 0.001 to 20.00 megohms (in 2 ranges)
- TDS Autoranging: 0.01 mg/L to 20.0 g/L (in 4 ranges)
- Temperature: -30° to +130°C (-22° to +266°F)
- Concentration: 0.000 to 9999 user units
- Salinity: 2 to 42 ppt
- Relative Accuracy: ±0.3%
- Types Automatic and manual, displays temperature °F/°C
- Reference Temperature (conductivity): References to international standard 25°C (77°F)
- Electronic Calibration Supplied Internal (1000 °mhos and 0.001 megohm)
- Data Output: RS-232-C, recorder jack
- Display: 0.5 in. digit LCD
- Net Weight: 1.5 lb. (0.7 kg)
- Dimensions (L x W x H): 8.25 x 6 x 3.5 in. (21 x 15.2 x 8.9 cm)
Order Info
Replacement glass probe (09-326-2A) and unbreakable epoxy plastic probe (22-278143) are sold separately.
Specifications
Specifications
| Includes | 115 V Power Adapter, 3 in. long (7.6 cm) Glass Probe, Receptacle to supply probe replatinizing current, 21 in. long (53.3 cm) Pivoting Probe Holder Arm, 9 V Battery |
| Accuracy (Conductivity) | ±0.3% |
| Outputs | RS232 |
| For Use With (Application) | Conductivity |
| Measurement Modes | Conductivity, Resistivity, Dissolved Solid, Salinity, Concentration, Temperature |
| Calibration Points | 4 |
| Range (Conductivity) | 0.01 μS to 200 mS |
| Range (Salinity) | 2 to 42 ppt |
| Range (TDS) | 0.01 mg/L to 20 g/L |
| Temperature Range (Metric) | -30°C to +130°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?