missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)
£29.60 - £923.28
Specifications
Content And Storage | Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area. |
---|---|
Form | Powder |
Grade | Molecular genetics |
Media Type | NZY Broth |
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
10122913
|
Fisher BioReagents™
BP2465-100 |
100 g | Poly Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11428443
|
Fisher BioReagents™
BP2465-2 |
2 kg | Poly Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10002864
|
Fisher BioReagents™
BP2465-10 |
10 kg | Poly Pail |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area. | |
Molecular genetics | |
Microbiology Media |
Powder | |
NZY Broth |