missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)
Specifications
Specifications
| Content And Storage | Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area. |
| Form | Powder |
| Grade | Molecular genetics |
| Media Type | NZY Broth |
| Product Type | Microbiology Media |
| Packaging Type | Poly Bottle |
| Quantity | 100 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?