Learn More
Spectrum Labs™ Fiber Filter Modules
£110.00 - £170.00
Specifications
Diameter (Metric) Inside | 0.5 mm |
---|---|
Filtration Area | 20 cm2 |
No. per Pack | 1 Each |
Material | Hollow Fiber |
Type | Filter Modules |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
12437061
|
Spectrum Labs™
C02-E100-05-N |
100 kDa |
£170.00
Each |
Log in to see stock. |
12437051
|
Spectrum Labs™
C02-E003-05-N |
3 kDa |
£110.00
Each |
Log in to see stock. |
Specifications
0.5 mm | |
1 Each | |
Filter Modules |
20 cm2 | |
Hollow Fiber |