Learn More
Spectrum Labs™ Fiber Filter Modules
Brand: Spectrum Labs™ C02-E003-05-N
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
Hollow Fiber | |
20 cm2 | |
1 Each | |
0.5 mm |
3 kDa | |
For Use with 1XFL mPES 3 Applications | |
Filter Modules |