Learn More
Spectrum Labs™ Fiber Filter Modules
Brand:  Spectrum Labs™ C02-E100-05-N
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12437061
  • £189.00 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Specifications
Hollow Fiber
20 cm2
1 Each
0.5 mm
100 kDa
For Use with 1XFL mPES Applications
Filter Modules