missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-4 Antibody (3A12G9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £416.00
Specifications
| Antigen | FGF-4 |
|---|---|
| Clone | 3A12G9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633756
|
Novus Biologicals
NBP2-37374-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18146098
|
Novus Biologicals
NBP2-37374 |
0.1 mL |
£416.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-4 Monoclonal specifically detects FGF-4 in Human samples. It is validated for Western Blot, ELISA.Specifications
| FGF-4 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
| FGF4 | |
| IgG1 | |
| Affinity Purified |
| 3A12G9 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| PBS with 0.05% Sodium Azide | |
| 2249 | |
| Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli. | |
| Primary | |
| 22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title