missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-4 Antibody (3A12G9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37374
This item is not returnable.
View return policy
Description
FGF-4 Monoclonal specifically detects FGF-4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| FGF-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
| Mouse | |
| 22 kDa | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 2249 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 3A12G9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FGF4 | |
| Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto