missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-4 Antibody (3A12G9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37374-0.025ml
This item is not returnable.
View return policy
Description
FGF-4 Monoclonal specifically detects FGF-4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| FGF-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
| Mouse | |
| 22 kDa | |
| 0.025 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 2249 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA | |
| 3A12G9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FGF4 | |
| Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction