Learn More
Epredia™ Lab Vision™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody.
£93.18 - £240.00
Specifications
| Antigen | alpha Fetoprotein (AFP) |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | IVD |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12663427
|
Epredia™
RB-9064-P0 |
100 μL | Concentrated | ||||||
|
12673427
|
Epredia™
RB-9064-P1 |
500 μL | Concentrated | ||||||
|
12653427
|
Epredia™
RB-9064-P |
1 mL | Concentrated | ||||||
|
12603437
|
Epredia™
RB-9064-R7 |
7 mL | Prediluted | ||||||
|
12613437
|
Epredia™
RB-9064-RQ |
12 mL | Ready-to-Use | ||||||
Description
AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Human α-fetoprotein purified from human cord serum
Molecular Weight: 70 kDa
Positive Control: HeLa cells, HepG2 cells, fetal liver or hepatocellular CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
| alpha Fetoprotein (AFP) | |
| Unconjugated | |
| IVD | |
| Human alpha-fetoprotein purified from human cord serum |
| Polyclonal | |
| Rabbit | |
| Oncogenesis | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.