missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody.

Brand:  Epredia™ RB-9064-RQ

 View more versions of this product

Product Code. 12613437

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Human α-fetoprotein purified from human cord serum

Molecular Weight: 70 kDa

Positive Control: HeLa cells, HepG2 cells, fetal liver or hepatocellular CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

alpha Fetoprotein (AFP)
Polyclonal
Rabbit
12 mL
Oncogenesis
Ready-to-Use
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Human alpha-fetoprotein purified from human cord serum
IVD
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.