missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Human α-fetoprotein purified from human cord serum
Molecular Weight: 70 kDa
Positive Control: HeLa cells, HepG2 cells, fetal liver or hepatocellular CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | alpha Fetoprotein (AFP) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Human alpha-fetoprotein purified from human cord serum |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Oncogenesis |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?