missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBPase-1 Inhibitor, Thermo Scientific™
A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1
£102.00 - £449.00
Specifications
Description | Off-white |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1 |
CAS | 883973-99-7 |
Molecular Formula | C13H7Cl3N2O3S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466819
|
Thermo Scientific Alfa Aesar
J65298.MF |
50 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15456819
|
Thermo Scientific Alfa Aesar
J65298.MA |
10 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Off-white | |
A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1 | |
C13H7Cl3N2O3S | |
F16BPase Inhibitor | |
Soluble in DMSO | |
Solid |
2°C to 8°C | |
883973-99-7 | |
377.60g/mol | |
Light Sensitive | |
Signal Transduction Reagent |