missing translation for 'onlineSavingsMsg'
Learn More

FBPase-1 Inhibitor, Thermo Scientific™

Product Code. 15456819
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 15456819

Brand: Thermo Scientific Alfa Aesar J65298.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description Off-white
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1
CAS 883973-99-7
Molecular Formula C13H7Cl3N2O3S
Formula Weight 377.60g/mol
Synonym F16BPase Inhibitor
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.