missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ TFP Ester-PEG4-DBCO

Product Code. 17621586 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mg
10 x 1 mg
Unit Size:
25mg
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17621586 10 x 1 mg Pack of 10
17514346 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17621586 Supplier Thermo Scientific™ Supplier No. C20043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An amine reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications. The crosslinker can be used to label a variety of amine containing molecules such as proteins, biomolecules, or small molecules.

  • An amine reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications
  • The crosslinker can be used to label a variety of amine containing molecules such as proteins, biomolecules, or small molecules
  • Once a protein or (bio)molecule is DBCO labeled, it can undergo a copper-free azide-alkyne cycloaddition reaction to form a stable triazole linkage with an azide labeled coupling partner
  • Can be used to create diverse conjugates with the biorthogonal coupling between an alkyne and azide (supplied separately)
  • The coupling or crosslinking procedure is typically done in two steps
    • First, a protein or amine containing molecule is labeled with a DBCO group and excess crosslinker is removed
    • Second, an azide containing moiety (prepared independently) is introduced into the same system as the DBCO moiety
    • Labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst
  • Ragent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
  • TFP esters are more hydrolytically stable than NHS esters
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Azide
Cleavable No
Description TFP Ester-PEG4-DBCO
Molecular Weight (g/mol) 714.7
PEGylated Yes
Spacer Arm Length 17.9 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability Not determined
Shipping Condition Wet Ice
Product Line EZ-Link No-Weigh
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety TFP Ester, DBCO
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 10 x 1 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.