missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips
Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips offer ultimate pipette and sample protection against unintentional, and unnoticed contamination
£46.75 - £185.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Certifications/Compliance | EN 1822 |
Format | Rack |
Product Line | ep DualFilter T.I.P.S |
Product Code | Brand | Length (Metric) | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Volume (Metric) | Price | Quantity & Availability | ||||
16472255
|
Eppendorf™
0030078500 |
34 mm | 0.1 to 10 μL |
£179.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16482255
|
Eppendorf™
0030078519 |
40 mm | 0.1 to 10 μL |
£179.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16492255
|
Eppendorf™
0030078527 |
46 mm | 0.5 to 20 μL |
£172.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16402265
|
Eppendorf™
0030078535 |
53 mm | 2 to 20 μL |
£172.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16412265
|
Eppendorf™
0030078543 |
53 mm | 2 to 100 μL |
£172.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16422265
|
Eppendorf™
0030078551 |
55 mm | 2 to 200 μL |
£172.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16432265
|
Eppendorf™
0030078560 |
55 mm | 20 to 300 μL |
£172.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16442265
|
Eppendorf™
0030078578 |
76 mm | 50 to 1000 μL |
£185.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
16452265
|
Eppendorf™
0030078594 |
103 mm | 50 to 1250 μL |
£95.05
Pack of 480 |
In Stock
Log in to see stock. |
||||
16455264
|
Eppendorf™
0030078586 |
115 mm | 0.25 to 2.5 mL |
£46.75
Pack of 240 |
In Stock
Log in to see stock. |
||||
16462265
|
Eppendorf™
0030078616 |
120 mm | 0.1 to 5 mL |
£93.15
Pack of 120 |
In Stock
Log in to see stock. |
||||
16472265
|
Eppendorf™
0030078624 |
175 mm | 0.2 to 5 mL |
£92.30
Pack of 120 |
In Stock
Log in to see stock. |
||||
10029212
|
Eppendorf™
0030077598 |
243 mm | 1000 to 10,000 μL |
£77.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips offer ultimate pipette and sample protection against unintentional, and unnoticed contamination
- A unique two-phase filter offers the ultimate protection of pipette, sample against unintentional and unnoticed contamination
- With defined flow dynamics, low wettability, and high thermal stability they are certified pyrogen-free and PCR clean
- Two filter layers provide dual protection against aerosols and biomolecules
- Free of PCR inhibiting additives provide for reliable downstream results
- Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
- Certificate of filter efficiency acc. to EN 1822
- Modern slim rack design with reclosable lid
- For use with: Air-cushion
- Improved sustainability with 20% less polypropylene consumed per rack
Specifications
Filtered | |
EN 1822 | |
ep DualFilter T.I.P.S |
Sterile | |
Rack | |
Polypropylene |