missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterobacter Cloacae Antibody (61/212) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £400.00
Specifications
| Antigen | Enterobacter Cloacae |
|---|---|
| Clone | 61/212 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18051636
|
Novus Biologicals
NB100-62544-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790294
|
Novus Biologicals
NB100-62544 |
0.1 mg |
£400.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enterobacter Cloacae Monoclonal specifically detects Enterobacter Cloacae in Bacteria samples. It is validated for ELISA.Specifications
| Enterobacter Cloacae | |
| 1.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| IgG2a | |
| Protein A purified | |
| This product recognizes Enterobacter cloacae, a gram-negative bacillus sometimes associated with urinary and respiratory tract infections in humans. |
| 61/212 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Enterobacter cloacae NCTC 10005 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title