missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterobacter Cloacae Antibody (61/212) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62544-0.025MG
This item is not returnable.
View return policy
Description
Enterobacter Cloacae Monoclonal specifically detects Enterobacter Cloacae in Bacteria samples. It is validated for ELISA.
Specifications
| Enterobacter Cloacae | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| This product recognizes Enterobacter cloacae, a gram-negative bacillus sometimes associated with urinary and respiratory tract infections in humans. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA | |
| 61/212 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Enterobacter cloacae NCTC 10005 | |
| 0.025 mg | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction