missing translation for 'onlineSavingsMsg'
Learn More

Elastin Antibody (BA-4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications

£175.00 - £453.00

Specifications

Antigen Elastin
Clone BA-4
Classification Monoclonal
Conjugate Unconjugated
Form Ascites
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18083074
View Documents
Novus Biologicals
NB100-2076-0.1ML
0.1 mL
£175.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18731494
View Documents
Novus Biologicals
NB100-2076
0.5 mL
£453.00
0.50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Elastin Monoclonal specifically detects Elastin in Human, Porcine, Canine, Feline, Guinea Pig, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Elastin
Monoclonal
Ascites
RUO
P15502
2006
Bovine alpha-Elastin.
Primary
Store at 4°C.
BA-4
Unconjugated
Mouse
Extracellular Matrix
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS
ELN
IgG1
Ascites
This recognizes insoluble elastin, alpha-elastin, soluble non-cross linked precursor of elastin (tropoelastin).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.