missing translation for 'onlineSavingsMsg'
Learn More

Elastin Antibody (BA-4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications

Brand:  Novus Biologicals NB100-2076-0.1ML

 View more versions of this product

Product Code. 18083074

  • £175.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Elastin Monoclonal specifically detects Elastin in Human, Porcine, Canine, Feline, Guinea Pig, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Elastin
Monoclonal
Unconjugated
Ascites, containing less than 0.1% sodium azide with Sodium Azide
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS
Mouse
Ascites
RUO
Primary
This recognizes insoluble elastin, alpha-elastin, soluble non-cross linked precursor of elastin (tropoelastin).
Store at 4°C.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
BA-4
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100
P15502
ELN
Bovine alpha-Elastin.
0.1 mL
Extracellular Matrix
2006
Guinea Pig, Feline, Pig, Canine, Sheep, Human, Goat
Ascites
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only