missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastin Antibody (BA-4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-2076
This item is not returnable.
View return policy
Description
Elastin Monoclonal antibody specifically detects Elastin in Human, Bovine, Feline, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| Elastin | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.05% Sodium Azide | |
| elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| This recognizes insoluble elastin, alpha-elastin, soluble non-cross linked precursor of elastin (tropoelastin). | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| BA-4 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:25-1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| P15502 | |
| ELN | |
| Bovine alpha-Elastin. | |
| 0.5 mL | |
| Extracellular Matrix | |
| 2006 | |
| Human, Bovine, Feline, Rabbit, Guinea Pig, Goat, Sheep | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction