missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGR1 Recombinant Rabbit Monoclonal Antibody (0Z9O1)

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen EGR1
Clone 0Z9O1
Concentration 1.08 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282549
View Documents
Invitrogen™
MA558095
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284154
View Documents
Invitrogen™
MA558096
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: ADTGEQPYEH LTAESFPDIS LNNEKVLVET SYPSQTTRLP PITYTGRFSL EPAPNSGNTL WPEPLFSLVS GLVSMTNPPA SSSSAPSPAA SSASASQSPP LSCAVPSNDS SPIYSAAPTF PTPNTDIFPE PQSQAFPGSA GTALQYPPPA YPAAKGGFQV P.

The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
Specifications

Specifications

EGR1
1.08 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P08046, P18146
13653, 1958
Recombinant fusion protein containing a sequence corresponding to amino acids 90-250 of human EGR1 (NP_001955.1).
Antibody
0Z9O1
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Human, Mouse
EGR1
A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225
EGR1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.