missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGR1 Recombinant Rabbit Monoclonal Antibody (0Z9O1)
Description
Immunogen sequence: ADTGEQPYEH LTAESFPDIS LNNEKVLVET SYPSQTTRLP PITYTGRFSL EPAPNSGNTL WPEPLFSLVS GLVSMTNPPA SSSSAPSPAA SSASASQSPP LSCAVPSNDS SPIYSAAPTF PTPNTDIFPE PQSQAFPGSA GTALQYPPPA YPAAKGGFQV P.
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
Specifications
Specifications
| Antigen | EGR1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 0Z9O1 |
| Concentration | 1.08 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | EGR1 |
| Gene Accession No. | P08046, P18146 |
| Gene Alias | A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?