missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGR1 Recombinant Rabbit Monoclonal Antibody (0Z9O1)

Product Code. 30282549
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30282549 20 μL 20µL
30284154 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30282549 Supplier Invitrogen™ Supplier No. MA558095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ADTGEQPYEH LTAESFPDIS LNNEKVLVET SYPSQTTRLP PITYTGRFSL EPAPNSGNTL WPEPLFSLVS GLVSMTNPPA SSSSAPSPAA SSASASQSPP LSCAVPSNDS SPIYSAAPTF PTPNTDIFPE PQSQAFPGSA GTALQYPPPA YPAAKGGFQV P.

The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 0Z9O1
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene EGR1
Gene Accession No. P08046, P18146
Gene Alias A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225
Gene Symbols EGR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-250 of human EGR1 (NP_001955.1).
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13653, 1958
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.