Learn More
Thermo Scientific™ DyLight™ 680-4xPEG Amine-Reactive Dye and Kits
Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.
Brand: Thermo Scientific™ 46603
Code : PN
Additional Details : Weight : 0.01000kg
Description
Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.
Additional Information and Online Ordering »
Highlights:
High fluorescence intensity – significantly brighter fluorescence than Alexa Fluor™ 680
PEGylated – improves solubility in aqueous solution, aids in cell permeability and improves tissue retention in in vivo applications
Convenient kit sizes – standard and microscale kit sizes to match your experimental needs
Recommended for:
- Fluorescence microscopy
- In vivo or ex vivo imaging
- Cell-based assays
- Flow cytometry/fluorescence-activated cell sorting (FACS)
Specifications
DyLight 680-4xPEG NHS Ester | |
NHS ester activated proprietary near-infrared fluorescent dye | |
5 x 65 μg | |
5 uses to label a total of 2mg of IgG using typical conditions |