Learn More
Thermo Scientific™ DyLight™ 680-4xPEG Amine-Reactive Dye and Kits
Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.
£196.00
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
13424219
|
Thermo Scientific™
46603 |
DyLight 680-4xPEG NHS Ester | 5 x 65 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.
Additional Information and Online Ordering »
Highlights:
High fluorescence intensity – significantly brighter fluorescence than Alexa Fluor™ 680
PEGylated – improves solubility in aqueous solution, aids in cell permeability and improves tissue retention in in vivo applications
Convenient kit sizes – standard and microscale kit sizes to match your experimental needs
Recommended for:
- Fluorescence microscopy
- In vivo or ex vivo imaging
- Cell-based assays
- Flow cytometry/fluorescence-activated cell sorting (FACS)