Learn More
DWK Life Sciences DURAN™ Replacement GLS 80™ Screw Cap Liner, PTFE Silicone
White PSU GLS 80™ wide-mouth screw cap (cap liner seal) and drip free, PTFE pouring ring for GLS 80™ bottles.
Brand:  DWK Life Sciences 1152921
Code : NEW
Additional Details : Weight : 0.09500kg
Product Code. 13258039
  • £37.70 / Pack of 5
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
The material used for this GLS 80 closure is a special compound based on polyarylsulphone. As a result, the chemical, thermal and mechanical properties of the material are improved compared to polypropylene. Thanks to the shortened triple thread, the DURAN™ GLS 80™ bottle can be opened or closed with only a three-quarter turn. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly sealed (cap liner: Platinum-cured silicone). A matching PTFE pouring ring is also available, permitting clean, drip-free use.
  • PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components
  • Screw cap seal with PTFE faced cap liner for efficient sealing
  • The matching PTFE pouring ring permits drip-free pouring and clean, safe working
Specifications
Specifications
Replacement cap liner
5/Pk.
79 mm
Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping
PSU, PBT, ETFE
GLS 80