missing translation for 'onlineSavingsMsg'
Learn More

DRP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DRP2
Concentration 0.4 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16885035
missing translation for 'viewDocuments'
Proteintech
26631-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
16875035
missing translation for 'viewDocuments'
Proteintech
26631-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Members of the dystrophin family of proteins perform a critical role in the maintece of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord.
Specifications

Specifications

DRP2
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q05AA6, Q13474, Q9EPA0
13497, 1821, 66027
DRP2 Fusion Protein Ag24462
Primary
-20°C
DRP2
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DRP 2, DRP2, dystrophin related protein 2
DRP2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.