missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26631-1-AP-20UL
This item is not returnable.
View return policy
Description
Members of the dystrophin family of proteins perform a critical role in the maintece of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord.Specifications
| DRP2 | |
| Polyclonal | |
| Unconjugated | |
| DRP2 | |
| DRP 2, DRP2, dystrophin related protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13497, 1821, 66027 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q05AA6, Q13474, Q9EPA0 | |
| DRP2 | |
| DRP2 Fusion Protein Ag24462 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion